accessibility menu, dialog, popup

UserName

OHBHO

CAS RN 5122-95-2

IUPAC Name: {[1,1'-biphenyl]-3-yl}boronic acid
Synonyms: [1,1'-biphenyl]-3-ylboronic acid
Molecular Weight (g/mol): 198.03
Molecular Formula: C12H11BO2
InChi Key: GOXICVKOZJFRMB-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results