CAS RN 51175-71-4
CAS RN 51175-71-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Thiophenesulfonyl chloride, ≥95%, Thermo Scientific™
CAS: 51175-71-4 Molecular Formula: C4H3ClO2S2 Molecular Weight (g/mol): 182.64 MDL Number: MFCD02682011 InChI Key: YSPWSQNKRBSICH-UHFFFAOYSA-N Synonym: 3-thiophenesulfonyl chloride,3-thiophenesulfonylchloride,chloro-3-thienylsulfone,acmc-209ksa,3-thienylsulfonyl chloride,3-thiophenesulphonylchloride,3-thienyl-sulfonyl chloride,3-chlorosulphonyl thiophene,3-thienyl sulfonyl chloride,3-thiophene sulfonyl chloride PubChem CID: 2776378 IUPAC Name: thiophene-3-sulfonyl chloride SMILES: ClS(=O)(=O)C1=CSC=C1