accessibility menu, dialog, popup

UserName

IH3CNCH3CH3H3C

CAS RN 51-93-4

IUPAC Name: ethyltrimethylazanium iodide
Synonyms: ethyltrimethylammonium iodide
Molecular Weight (g/mol): 215.08
Molecular Formula: C5H14IN
InChi Key: ZPEBBUBSCOELHI-UHFFFAOYSA-M
SMILES: [I-].CC[N+](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results