accessibility menu, dialog, popup

UserName

CAS RN 50990-93-7

CH3OCH3ClO

CAS RN 50990-93-7

IUPAC Name: 2,5-dimethylfuran-3-carbonyl chloride
Synonyms: 2,5-dimethylfuran-3-carbonyl chloride
Molecular Weight (g/mol): 158.58
Molecular Formula: C7H7ClO2
InChi Key: RWORXZHVOUPMMM-UHFFFAOYSA-N
SMILES: CC1=CC(C(Cl)=O)=C(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)