accessibility menu, dialog, popup

UserName

ONSOOO

CAS RN 50978-45-5

IUPAC Name: 1,1,3-trioxo-2,3-dihydro-1λ⁶,2-benzothiazole-2-carbaldehyde
Synonyms: 1,1,3-trioxo-1λ⁶,2-benzothiazole-2-carbaldehyde
Molecular Weight (g/mol): 211.19
Molecular Formula: C8H5NO4S
InChi Key: LMPIFZSJKLLOLM-UHFFFAOYSA-N
SMILES: O=CN1C(=O)C2=CC=CC=C2S1(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results