accessibility menu, dialog, popup

UserName

CAS RN 50595-15-8

CH3CH3H3COOHO

CAS RN 50595-15-8

IUPAC Name: tert-butyl 2-hydroxyacetate
Synonyms: tert-butyl 2-hydroxyacetate
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: WINGEFIITRDOLJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results