accessibility menu, dialog, popup

UserName

CAS RN 50501-07-0

H3COOHN

CAS RN 50501-07-0

IUPAC Name: ethyl 2,3-dihydro-1H-indole-2-carboxylate
Synonyms: ethyl 2,3-dihydro-1H-indole-2-carboxylate
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: KISPUTPAKVZNBI-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CC2=CC=CC=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results