accessibility menu, dialog, popup

UserName

H3CNCH3H3CH3CClAuCl

CAS RN 50480-99-4

IUPAC Name: dichlorogolduide; tetrabutylazanium
Synonyms: dichlorogolduide; tetrabutylammonium ion
Molecular Weight (g/mol): 510.34
Molecular Formula: C16H36AuCl2N
InChi Key: XZONJCOZEGOEDY-UHFFFAOYSA-L
SMILES: Cl[Au-]Cl.CCCC[N+](CCCC)(CCCC)CCCC
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results