accessibility menu, dialog, popup

UserName

CH3NHOCH3HNOONH2O

CAS RN 50444-54-7

IUPAC Name: benzyl N-{1-[(1-carbamoylethyl)carbamoyl]ethyl}carbamate
Synonyms: benzyl N-{1-[(1-carbamoylethyl)carbamoyl]ethyl}carbamate
Molecular Weight (g/mol): 293.32
Molecular Formula: C14H19N3O4
InChi Key: WEIOJLPDGBBVCH-UHFFFAOYNA-N
SMILES: CC(NC(=O)C(C)NC(=O)OCC1=CC=CC=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)