accessibility menu, dialog, popup

UserName

FN

CAS RN 503-20-8

IUPAC Name: 2-fluoroacetonitrile
Synonyms: acetonitrile, fluoro-
Molecular Weight (g/mol): 59.04
Molecular Formula: C2H2FN
InChi Key: GNFVFPBRMLIKIM-UHFFFAOYSA-N
SMILES: FCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results