accessibility menu, dialog, popup

UserName

CAS RN 49678-04-8

IUPAC Name: 3-(4-bromophenyl)prop-2-enal
Synonyms: 3-(4-bromophenyl)prop-2-enal
Molecular Weight (g/mol): 211.06
Molecular Formula: C9H7BrO
InChi Key: XYRAWLRFGKLUMW-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=CC=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results