CAS RN 49619-58-1
IUPAC Name:
4-oxo-6-(propan-2-yl)-4H-chromene-3-carbaldehyde
Synonyms:
6-isopropyl-4-oxochromene-3-carbaldehyde
Molecular Weight (g/mol):
216.24
Molecular Formula:
C13H12O3
InChi Key:
FRRYMYQANNFABF-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=C2OC=C(C=O)C(=O)C2=C1
1
–
1
of
1
results
1
3-Formyl-6-isopropylchromone 98.0+%, TCI America™
CAS: 49619-58-1 Molecular Formula: C13H12O3 Molecular Weight (g/mol): 216.24 MDL Number: MFCD00192183 InChI Key: FRRYMYQANNFABF-UHFFFAOYSA-N Synonym: 6-Isopropyl-4-oxo-4H-1-benzopyran-3-carboxaldehyde, 6-Isopropylchromone-3-carboxaldehyde PubChem CID: 688711 IUPAC Name: 4-oxo-6-(propan-2-yl)-4H-chromene-3-carbaldehyde SMILES: CC(C)C1=CC=C2OC=C(C=O)C(=O)C2=C1