accessibility menu, dialog, popup

UserName

CAS RN 4940-11-8

IUPAC Name: 2-ethyl-3-hydroxy-4H-pyran-4-one
Synonyms: ethyl maltol ethyl hydroxypyrone
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: YIKYNHJUKRTCJL-UHFFFAOYSA-N
SMILES: CCC1=C(O)C(=O)C=CO1
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)