CAS RN 4857-81-2
CAS RN 4857-81-2
IUPAC Name:
(2-chlorophenyl)bis(4-{ethyl[(3-sulfophenyl)methyl]amino}phenyl)methylium
Synonyms:
(2-chlorophenyl)bis(4-{ethyl[(3-sulfophenyl)methyl]amino}phenyl)methylium
Molecular Weight (g/mol):
704.27
Molecular Formula:
C37H36ClN2O6S2
InChi Key:
KZMRYBLIGYQPPP-UHFFFAOYSA-O
SMILES:
CCN(CC1=CC=CC(=C1)S(O)(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S(O)(=O)=O)C1=CC=CC=C1Cl
1
–
1
of
1
results
1
Acid Green 9, TCI America™
CAS: 4857-81-2 Molecular Formula: C37H34ClN2NaO6S2 Molecular Weight (g/mol): 725.247 MDL Number: MFCD00050440 InChI Key: SHBDDIJUSNNBLQ-UHFFFAOYSA-M Synonym: Patent Green, Alizarin Green, Brilliant Acid Green, Brilliant Milling Green PubChem CID: 25021982 IUPAC Name: sodium;3-[[4-[(2-chlorophenyl)-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate SMILES: CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5Cl.[Na+]