accessibility menu, dialog, popup

UserName

ONMgBr

CAS RN 480424-77-9

IUPAC Name: 4-{[2-(bromomagnesio)phenyl]methyl}morpholine
Synonyms: 4-{[2-(bromomagnesio)phenyl]methyl}morpholine
Molecular Weight (g/mol): 280.45
Molecular Formula: C11H14BrMgNO
InChi Key: RSBWIKCTZYDPJQ-UHFFFAOYSA-M
SMILES: Br[Mg]C1=C(CN2CCOCC2)C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results