accessibility menu, dialog, popup

UserName

CAS RN 47729-63-5

IUPAC Name: 5,6-dichloro-2-[3-(5,6-dichloro-1,3-diethyl-2,3-dihydro-1H-1,3-benzodiazol-2-ylidene)prop-1-en-1-yl]-1,3-diethyl-1H-1,3-benzodiazol-3-ium iodide
Synonyms: JC1 dye
Molecular Weight (g/mol): 652.22
Molecular Formula: C25H27Cl4IN4
InChi Key: FYNNIUVBDKICAX-UHFFFAOYSA-M
SMILES: [I-].CCN1C(=CC=CC2=[N+](CC)C3=CC(Cl)=C(Cl)C=C3N2CC)N(CC)C2=CC(Cl)=C(Cl)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results