accessibility menu, dialog, popup

UserName

OSb

CAS RN 4756-75-6

IUPAC Name: triphenylstibanone
Synonyms: stibine oxide, triphenyl-
Molecular Weight (g/mol): 369.08
Molecular Formula: C18H15OSb
InChi Key: RIBJLXBLWQKWLO-UHFFFAOYSA-N
SMILES: O=[Sb](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)