accessibility menu, dialog, popup

UserName

CAS RN 4706-17-6

OHPOHOH

CAS RN 4706-17-6

IUPAC Name: 3-[bis(3-hydroxypropyl)phosphanyl]propan-1-ol
Synonyms: 3-[bis(3-hydroxypropyl)phosphanyl]propan-1-ol
Molecular Weight (g/mol): 208.24
Molecular Formula: C9H21O3P
InChi Key: YICAEXQYKBMDNH-UHFFFAOYSA-N
SMILES: OCCCP(CCCO)CCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results