CAS RN 4680-78-8
CAS RN 4680-78-8
IUPAC Name:
sodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(phenyl)methylium
Synonyms:
sodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(phenyl)methylium
Molecular Weight (g/mol):
690.8
Molecular Formula:
C37H35N2NaO6S2
InChi Key:
XKTMIJODWOEBKO-UHFFFAOYSA-M
SMILES:
[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O
1
–
1
of
1
results
1
Acid Green 3, TCI America™
CAS: 4680-78-8 Molecular Formula: C37H35N2NaO6S2 Molecular Weight (g/mol): 690.805 MDL Number: MFCD00012122 InChI Key: XKTMIJODWOEBKO-UHFFFAOYSA-M Synonym: Acid Green B, Guinea Green B, Pontacyl Green B PubChem CID: 20803 IUPAC Name: sodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-phenylmethyl]anilino]methyl]benzenesulfonate SMILES: CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5.[Na+]