CAS RN 46006-36-4
IUPAC Name:
1H-1,3-benzodiazole-4-carboxylic acid
Synonyms:
1H-1,3-benzodiazole-4-carboxylic acid
Molecular Weight (g/mol):
162.15
Molecular Formula:
C8H6N2O2
InChi Key:
VVQNAFBGAWCMLU-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C2N=CNC2=CC=C1
1
–
1
of
1
results
1
1H-Benzimidazole-4-carboxylic acid, 97%
CAS: 46006-36-4 Molecular Formula: C8H6N2O2 Molecular Weight (g/mol): 162.15 MDL Number: MFCD01823426 InChI Key: VVQNAFBGAWCMLU-UHFFFAOYSA-N Synonym: 1h-benzoimidazole-4-carboxylic acid,1h-benzimidazole-7-carboxylic acid,benzimidazole-4-carboxylic acid,1h-benzo d imidazole-4-carboxylic acid,1h-1,3-benzodiazole-4-carboxylic acid,1h-benzo d imidazole-7-carboxylic acid,3h-1,3-benzodiazole-4-carboxylic acid,1h-benzimidazole-4-carboxylicacid,acmc-1aqhf,4-carboxy-1h-benzimidazole PubChem CID: 2771758 SMILES: OC(=O)C1=C2N=CNC2=CC=C1