accessibility menu, dialog, popup

UserName

H3COOHHO(E)

CAS RN 458-35-5

IUPAC Name: 4-[(1E)-3-hydroxyprop-1-en-1-yl]-2-methoxyphenol
Synonyms: coniferyl alcohol coniferol
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: JMFRWRFFLBVWSI-NSCUHMNNSA-N
SMILES: COC1=CC(\C=C\CO)=CC=C1O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)