accessibility menu, dialog, popup

UserName

H3CSiH3CCH3S

CAS RN 4551-15-9

IUPAC Name: trimethyl(phenylsulfanyl)silane
Synonyms: trimethyl(phenylsulfanyl)silane
Molecular Weight (g/mol): 182.36
Molecular Formula: C9H14SSi
InChi Key: VJMQFIRIMMSSRW-UHFFFAOYSA-N
SMILES: C[Si](C)(C)SC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results