accessibility menu, dialog, popup

UserName

CAS RN 45125-00-6

H3CCH3H3COOOOHNH2

CAS RN 45125-00-6

IUPAC Name: 2-amino-5-(tert-butoxy)-5-oxopentanoic acid
Synonyms: 2-amino-5-(tert-butoxy)-5-oxopentanoic acid
Molecular Weight (g/mol): 203.24
Molecular Formula: C9H17NO4
InChi Key: OIOAKXPMBIZAHL-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CCC(N)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)