accessibility menu, dialog, popup

UserName

OFFFCl

CAS RN 450-96-4

IUPAC Name: 1-chloro-2-(trifluoromethoxy)benzene
Synonyms: 1-chloro-2-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 196.55
Molecular Formula: C7H4ClF3O
InChi Key: NZRLCYJRHKUVCV-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results