accessibility menu, dialog, popup

UserName

NClNOOCl

CAS RN 4487-56-3

IUPAC Name: 2,4-dichloro-5-nitropyridine
Synonyms: 2,4-dichloro-5-nitropyridine
Molecular Weight (g/mol): 192.98
Molecular Formula: C5H2Cl2N2O2
InChi Key: RZVJQUMDJUUBBF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CN=C(Cl)C=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results