accessibility menu, dialog, popup

UserName

CAS RN 447-53-0

IUPAC Name: 1,2-dihydronaphthalene
Synonyms: dialin delta(1)-dialin diolin delta1-dialin 1,2-dialin
Molecular Weight (g/mol): 130.19
Molecular Formula: C10H10
InChi Key: KEIFWROAQVVDBN-UHFFFAOYSA-N
SMILES: C1CC2=CC=CC=C2C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results