accessibility menu, dialog, popup

UserName

CH3OOOHOHOHOHONH2NCH3H3CHCl

CAS RN 4465-65-0

IUPAC Name: 4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-3,4,4a,5,12,12a-hexahydrotetracene-2-carboxamide hydrochloride
Synonyms: 4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide hydrochloride
Molecular Weight (g/mol): 462.88
Molecular Formula: C22H23ClN2O7
InChi Key: SPFAOPCHYIJPHJ-UHFFFAOYNA-N
SMILES: Cl.CN(C)C1C2CC3=C(C(O)=C4C(O)=CC=CC4=C3C)C(=O)C2(O)C(O)=C(C(N)=O)C1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)