accessibility menu, dialog, popup

UserName

CAS RN 4463-44-9

HNaCH3NHH3CNCH3CH3OHSOHOOSHOOO(Z)(E/Z)

CAS RN 4463-44-9

IUPAC Name: sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-6-hydroxybenzene-1,3-disulfonic acid hydride
Synonyms: sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-6-hydroxybenzene-1,3-disulfonic acid hydride
Molecular Weight (g/mol): 556.62
Molecular Formula: C25H29N2NaO7S2
InChi Key: HLEQELPTFNOLFE-ZCTWALFCSA-N
SMILES: [H-].[Na+].CCNC1=CC=C(C=C1C)C(=C1/C=CC(=NCC)C(C)=C1)\C1=CC(O)=C(C=C1S(O)(=O)=O)S(O)(=O)=O
Color 
  • (5)
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)