accessibility menu, dialog, popup

UserName

BOB

CAS RN 4426-21-5

IUPAC Name: diphenylboranyl diphenylborinate
Synonyms: diphenylboranyl diphenylborinate
Molecular Weight (g/mol): 346.04
Molecular Formula: C24H20B2O
InChi Key: SNQFEECGHGUHBK-UHFFFAOYSA-N
SMILES: O(B(C1=CC=CC=C1)C1=CC=CC=C1)B(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results