CAS RN 442517-33-1
IUPAC Name:
6-ethynylquinoxaline
Synonyms:
6-ethynylquinoxaline
Molecular Weight (g/mol):
154.17
Molecular Formula:
C10H6N2
InChi Key:
QSYDWUWZZBZOCD-UHFFFAOYSA-N
SMILES:
C#CC1=CC=C2N=CC=NC2=C1
1
–
1
of
1
results
1
6-Ethynylquinoxaline, ≥97%, Thermo Scientific™
CAS: 442517-33-1 Molecular Formula: C10H6N2 Molecular Weight (g/mol): 154.172 MDL Number: MFCD08435845 InChI Key: QSYDWUWZZBZOCD-UHFFFAOYSA-N Synonym: quinoxaline, 6-ethynyl,quinoxaline,6-ethynyl,quinoxalin-6-yl acetylene PubChem CID: 18525704 IUPAC Name: 6-ethynylquinoxaline SMILES: C#CC1=CC2=NC=CN=C2C=C1