accessibility menu, dialog, popup

UserName

CAS RN 4402-32-8

H3CNH3CCH3OH

CAS RN 4402-32-8

IUPAC Name: 1-(diethylamino)propan-2-ol
Synonyms: 1-(diethylamino)propan-2-ol
Molecular Weight (g/mol): 131.22
Molecular Formula: C7H17NO
InChi Key: BHUXAQIVYLDUQV-UHFFFAOYNA-N
SMILES: CCN(CC)CC(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results