accessibility menu, dialog, popup

UserName

CAS RN 440122-66-7

IUPAC Name: 4-(7-bromo-5-hydroxy-2,3-dihydro-1,3-benzoxazol-2-ylidene)cyclohexa-2,5-dien-1-one
Synonyms: 4-(7-bromo-5-hydroxy-3H-1,3-benzoxazol-2-ylidene)cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 306.12
Molecular Formula: C13H8BrNO3
InChi Key: IVFWABSHZFOMIG-UHFFFAOYSA-N
SMILES: OC1=CC(Br)=C2OC(NC2=C1)=C1C=CC(=O)C=C1
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results