accessibility menu, dialog, popup

UserName

CAS RN 4395-87-3

IUPAC Name: 2-[4-(propan-2-yl)phenyl]acetonitrile
Synonyms: 2-(4-isopropylphenyl)acetonitrile
Molecular Weight (g/mol): 159.23
Molecular Formula: C11H13N
InChi Key: RIPHZOPMCRSGSI-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(CC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results