accessibility menu, dialog, popup

UserName

OHBrOBrOOH

CAS RN 4370-59-6

IUPAC Name: 2,5-dibromo-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione
Synonyms: 2,5-dibromo-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol): 297.89
Molecular Formula: C6H2Br2O4
InChi Key: GMZWPTALVQRAFV-UHFFFAOYSA-N
SMILES: OC1=C(Br)C(=O)C(O)=C(Br)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results