accessibility menu, dialog, popup

UserName

OHOHO

CAS RN 4358-64-9

IUPAC Name: oxolane-3,4-diol
Synonyms: oxolane-3,4-diol
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: SSYDTHANSGMJTP-UHFFFAOYNA-N
SMILES: OC1COCC1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results