accessibility menu, dialog, popup

UserName

CAS RN 4332-95-0

OHNHSNO

CAS RN 4332-95-0

IUPAC Name: 5-[(4-hydroxyphenyl)methyl]-3-phenyl-2-sulfanylideneimidazolidin-4-one
Synonyms: 5-[(4-hydroxyphenyl)methyl]-3-phenyl-2-sulfanylideneimidazolidin-4-one
Molecular Weight (g/mol): 298.36
Molecular Formula: C16H14N2O2S
InChi Key: LCFDDJINTNHAEQ-UHFFFAOYNA-N
SMILES: OC1=CC=C(CC2NC(=S)N(C2=O)C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)