accessibility menu, dialog, popup

UserName

CAS RN 4313-03-5

CH3O(E)(E)

CAS RN 4313-03-5

IUPAC Name: (2E,4E)-hepta-2,4-dienal
Synonyms: 2,4-heptadienal, (E,E)- heptadienal
Molecular Weight (g/mol): 110.16
Molecular Formula: C7H10O
InChi Key: SATICYYAWWYRAM-VNKDHWASSA-N
SMILES: CC\C=C\C=C\C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results