CAS RN 4306-65-4
CAS RN 4306-65-4
IUPAC Name:
1,1,3,5-tetramethylcyclohexane
Synonyms:
1,1,3,5-tetramethylcyclohexane
Molecular Weight (g/mol):
140.27
Molecular Formula:
C10H20
InChi Key:
WOJSMJIXPQLESQ-UHFFFAOYNA-N
SMILES:
CC1CC(C)CC(C)(C)C1
1
–
1
of
1
results
1
1,1,3,5-Tetramethylcyclohexane 96.0+%, TCI America™
CAS: 4306-65-4 Molecular Formula: C10H20 Molecular Weight (g/mol): 140.27 MDL Number: MFCD00060820 InChI Key: WOJSMJIXPQLESQ-UHFFFAOYSA-N PubChem CID: 107263 IUPAC Name: 1,1,3,5-tetramethylcyclohexane SMILES: CC1CC(CC(C1)(C)C)C