accessibility menu, dialog, popup

UserName

CAS RN 4261-68-1

IUPAC Name: hydrogen (2-chloroethyl)bis(propan-2-yl)amine chloride
Synonyms: hydrogen (2-chloroethyl)diisopropylamine chloride
Molecular Weight (g/mol): 200.15
Molecular Formula: C8H19Cl2N
InChi Key: IUSXYVRFJVAVOB-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)N(CCCl)C(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results