accessibility menu, dialog, popup

UserName

H3COHO

CAS RN 4250-38-8

IUPAC Name: nonacosanoic acid
Synonyms: nonacosanoic acid
Molecular Weight (g/mol): 438.78
Molecular Formula: C29H58O2
InChi Key: IHEJEKZAKSNRLY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results