accessibility menu, dialog, popup

UserName

CAS RN 42182-27-4

NH2NN

CAS RN 42182-27-4

IUPAC Name: 2-aminopyridine-4-carbonitrile
Synonyms: 2-aminopyridine-4-carbonitrile
Molecular Weight (g/mol): 119.13
Molecular Formula: C6H5N3
InChi Key: GEEAYLFEIFJFGP-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=N1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results