accessibility menu, dialog, popup

UserName

OEuOOOOOCHCHHCFFFFFFFFFSSS

CAS RN 41884-77-9

IUPAC Name: europium(3+) tris(4,4,4-trifluoro-1,3-dioxo-1-(thiophen-2-yl)butan-2-ide)
Synonyms: europium(3+) tris(4,4,4-trifluoro-1,3-dioxo-1-(thiophen-2-yl)butan-2-ide)
Molecular Weight (g/mol): 815.48
Molecular Formula: C24H12EuF9O6S3
InChi Key: JIYALKNSIOHINA-UHFFFAOYSA-N
SMILES: [Eu+3].FC(F)(F)C(=O)[CH-]C(=O)C1=CC=CS1.FC(F)(F)C(=O)[CH-]C(=O)C1=CC=CS1.FC(F)(F)C(=O)[CH-]C(=O)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results