accessibility menu, dialog, popup

UserName

CH3H3CCH3O(S)O(R)OO(R)HO(S)HO(S)OH(R)OH(S)HO(S)HO(R)OH(S)(R)CH3(R)(R)HO(R)(R)H3C(S)O(R)O(R)OH(S)OH(S)OH(R)O(S)O(R)HO(S)OH(S)OH(R)HOCH3H3C(R)(R)H3C

CAS RN 41753-43-9

IUPAC Name: (2S,3R,4S,5S,6R)-2-({2-[(1S,3aR,3bR,5aR,7S,9aR,9bR,11R,11aR)-7-{[(2R,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]oxy}-11-hydroxy-3a,3b,6,6,9a-pentamethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-6-methylhept-5-en-2-yl}oxy)-6-({[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxane-3,4,5-triol
Synonyms: (2S,3R,4S,5S,6R)-2-({2-[(1S,3aR,3bR,5aR,7S,9aR,9bR,11R,11aR)-7-{[(2R,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]oxy}-11-hydroxy-3a,3b,6,6,9a-pentamethyl-dodecahydro-1H-cyclopenta[a]phenanthren-1-yl]-6-methylhept-5-en-2-yl}oxy)-6-({[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 1109.31
Molecular Formula: C54H92O23
InChi Key: GZYPWOGIYAIIPV-NGBMAODDSA-N
SMILES: CC(C)=CCCC(C)(O[C@@H]1O[C@H](COC2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@H](O)[C@H]1O)[C@H]1CC[C@]2(C)[C@@H]1[C@H](O)C[C@@H]1[C@@]3(C)CC[C@H](O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)C(C)(C)[C@@H]3CC[C@@]21C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)