accessibility menu, dialog, popup

UserName

NSClCl

CAS RN 4175-76-2

IUPAC Name: 2,4-dichloro-1,3-thiazole
Synonyms: 2,4-dichloro-1,3-thiazole
Molecular Weight (g/mol): 154.01
Molecular Formula: C3HCl2NS
InChi Key: ICETWLGKJXCIDX-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results