accessibility menu, dialog, popup

UserName

CH3(R)HN

CAS RN 41720-98-3

IUPAC Name: (2R)-2-methylpyrrolidine
Synonyms: (2R)-2-methylpyrrolidine
Molecular Weight (g/mol): 85.15
Molecular Formula: C5H11N
InChi Key: RGHPCLZJAFCTIK-RXMQYKEDSA-N
SMILES: C[C@@H]1CCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results