accessibility menu, dialog, popup

UserName

HPBH3

CAS RN 41593-58-2

IUPAC Name: diphenylphosphane borane
Synonyms: diphenylphosphine borane
Molecular Weight (g/mol): 200.03
Molecular Formula: C12H14BP
InChi Key: XUFGOLYVDOBYLK-UHFFFAOYSA-N
SMILES: B.P(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)