accessibility menu, dialog, popup

UserName

SHSH

CAS RN 41563-69-3

IUPAC Name: [3-(sulfanylmethyl)phenyl]methanethiol
Synonyms: [3-(sulfanylmethyl)phenyl]methanethiol
Molecular Weight (g/mol): 170.29
Molecular Formula: C8H10S2
InChi Key: JSNABGZJVWSNOB-UHFFFAOYSA-N
SMILES: SCC1=CC(CS)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results