accessibility menu, dialog, popup

UserName

CH3OOO

CAS RN 41302-34-5

IUPAC Name: methyl 2-oxocyclohexane-1-carboxylate
Synonyms: methyl 2-oxocyclohexane-1-carboxylate
Molecular Weight (g/mol): 156.18
Molecular Formula: C8H12O3
InChi Key: JEENWEAPRWGXSG-UHFFFAOYNA-N
SMILES: COC(=O)C1CCCCC1=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)