accessibility menu, dialog, popup

UserName

CAS RN 4119-41-9

IUPAC Name: (3-iodopropyl)benzene
Synonyms: G-phenyl propyliodine
Molecular Weight (g/mol): 246.09
Molecular Formula: C9H11I
InChi Key: RGCKJSPKMTWLLX-UHFFFAOYSA-N
SMILES: ICCCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)