CAS RN 411206-75-2
IUPAC Name:
potassium (3-chlorophenyl)trifluoroboranuide
Synonyms:
potassium (3-chlorophenyl)trifluoroboranuide
Molecular Weight (g/mol):
218.45
Molecular Formula:
C6H4BClF3K
InChi Key:
LRJCVRYGKNDLAD-UHFFFAOYSA-N
SMILES:
[K+].F[B-](F)(F)C1=CC(Cl)=CC=C1
1
–
1
of
1
results
1
Promotions Available
Potassium 3-chlorophenyltrifluoroborate, 96%, Thermo Scientific™
Potassium 3-chlorophenyltrifluoroborate, 96%, Thermo Scientific™
CAS: 411206-75-2 Molecular Formula: C6H4BClF3K Molecular Weight (g/mol): 218.452 MDL Number: MFCD04115754 InChI Key: LRJCVRYGKNDLAD-UHFFFAOYSA-N Synonym: potassium 3-chlorophenyl trifluoroborate,potassium 3-chlorophenyl trifluoroboranuide,potassium 3-chlorophenyltrifluoroborate,amtb707,potassium 3-chlorophenyl-trifluoroboranuide,potassium 3-chlorophenyl trifluoro borate 1-,potassium ion 3-chlorophenyl trifluoroboranuide,potassium 3-chlorophenyl-tris fluoranyl boranuide PubChem CID: 23677818 IUPAC Name: potassium;(3-chlorophenyl)-trifluoroboranuide SMILES: [B-](C1=CC(=CC=C1)Cl)(F)(F)F.[K+]